Metamizol-Natriummonohydrat
Thermo Scientific Chemicals

Metamizol-Natriummonohydrat

Have Questions?
Ansicht ändernbuttonViewtableView
KatalognummerMenge
4674800101 g
467482500250 mg
Katalognummer 467480010
Preis (EUR)
210,00
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
210,00
Each
Chemikalien-Kennzeichnungen
CAS5907-38-0
IUPAC Namesodium [(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)(methyl)amino]methanesulfonate hydrate
Molecular FormulaC13H18N3NaO5S
InChI KeyUNZIDPIPYUMVPA-UHFFFAOYSA-M
SMILESO.[Na+].CN(CS([O-])(=O)=O)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
HPLC>=97.5 %
Melting point211°C to 216°C
Water4.5 to 5.5 %
Appearance (Form)Powder or crystals
Appearance (Color)White to light yellow to light beige
Mehr anzeigen
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only