3-Ciclopenten-1-ol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Ciclopenten-1-ol, Thermo Scientific Chemicals

CAS: 14320-38-8 | C5H8O | 84.12 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Número de catálogo 467550010
Precio (EUR)
68,20
Each
Cantidad:
1 g
Pedido a granel o personalizado
Precio (EUR)
68,20
Each
Identificadores químicos
CAS14320-38-8
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Liquid
Water=<1 %
Appearance (Color)Colorless to yellow to brown
GC>=94 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only