Sodium thiomethoxide, 90+%, technical
Thermo Scientific Chemicals

Sodium thiomethoxide, 90+%, technical

CAS: 5188-07-8 | CH3NaS | 70.08 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
產品號碼 469890010
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS5188-07-8
IUPAC Namesodium methylsulfanide
Molecular FormulaCH3NaS
InChI KeyRMBAVIFYHOYIFM-UHFFFAOYSA-M
SMILES[Na+].C[S-]
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to light yellow to light brown
Titration Iodimetric>=90.0 %
Appearance (Form)Powder or granular powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only