CD3 Monoclonal Antibody (17A2), APC-eFluor™ 780, eBioscience™ - FAQs
CD3 Monoclonal Antibody (17A2), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (17A2), APC-eFluor™ 780, eBioscience™ - FAQs (47-0032-80, 47-0032-82)