CD3 Monoclonal Antibody (SK7), APC-eFluor™ 780, eBioscience™ - FAQs
CD3 Monoclonal Antibody (SK7), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (SK7), APC-eFluor™ 780, eBioscience™ - FAQs (47-0036-42, 47-0036-41)