CD8a Monoclonal Antibody (SK1), APC-eFluor™ 780, eBioscience™ - FAQs
CD8a Monoclonal Antibody (SK1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (SK1), APC-eFluor™ 780, eBioscience™ - FAQs (47-0087-42, 47-0087-41)