CD44 Monoclonal Antibody (IM7), APC-eFluor™ 780, eBioscience™ - FAQs
CD44 Monoclonal Antibody (IM7), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD44 Monoclonal Antibody (IM7), APC-eFluor™ 780, eBioscience™ - FAQs (47-0441-82, 47-0441-80)