CD45.1 Monoclonal Antibody (A20), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD45.1 Monoclonal Antibody (A20), APC-eFluor™ 780, eBioscience™ - FAQs (47-0453-80, 47-0453-82)