CD45RO Monoclonal Antibody (UCHL1), APC-eFluor™ 780, eBioscience™ - FAQs
CD45RO Monoclonal Antibody (UCHL1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD45RO Monoclonal Antibody (UCHL1), APC-eFluor™ 780, eBioscience™ - FAQs (47-0457-42, 47-0457-41)