CD64 Monoclonal Antibody (X54-5/7.1), APC-eFluor™ 780, eBioscience™ - FAQs
CD64 Monoclonal Antibody (X54-5/7.1), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for CD64 Monoclonal Antibody (X54-5/7.1), APC-eFluor™ 780, eBioscience™ - FAQs (47-0641-80, 47-0641-82)