EOMES Monoclonal Antibody (WD1928), APC-eFluor™ 780, eBioscience™ - FAQs
EOMES Monoclonal Antibody (WD1928), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for EOMES Monoclonal Antibody (WD1928), APC-eFluor™ 780, eBioscience™ - FAQs (47-4877-42, 47-4877-41)