NK1.1 Monoclonal Antibody (PK136), APC-eFluor™ 780, eBioscience™ - FAQs
NK1.1 Monoclonal Antibody (PK136), APC-eFluor™ 780, eBioscience™ - FAQs
View additional product information for NK1.1 Monoclonal Antibody (PK136), APC-eFluor™ 780, eBioscience™ - FAQs (47-5941-82, 47-5941-80)