B*14 SSP UniTray® Kit with Taq DNA Polymerase
Catalog number: 470912
 Related applications: Diagnostic Testing
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
• For use when high resolution typing of HLA Class I is required and low resolution results are known.
• Provide allele-level typing results with minimal ambiguities.
• Pre-dispensed liquid primer mixes
• Includes pre-aliquoted amplification buffer
• Compatible with UniMatch Plus Analysis software
• Allele-updating availabe via the web
Contents & storage
Store in freezer (-5 to -30°C).
Documents
Manuals & protocols