Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Immuno Standard Modules White and Black
Thermo Scientific™

Immuno Standard Modules White and Black

Conduct fluorescence- and luminescence-based immunoassays and binding assays using these immuno standard modules white and black.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorVolume (Metric) WellSurface Treatment
475523Black400 μLPolySorp
437591White350 μLMaxiSorp
437702White350 μLPolySorp
437915Clear350 μLMaxiSorp
475515Black400 μLMaxiSorp
7566White380 μLHydrophobic
Catalog number 475523
Price (USD)
627.57
Online exclusive
731.82
Save 104.25
Case of 60
Color:
Black
Volume (Metric) Well:
400 μL
Surface Treatment:
PolySorp
Price (USD)
627.57
Online exclusive
731.82
Save 104.25
Case of 60
Conduct fluorescence- and luminescence-based immunoassays and binding assays using Thermo Scientific™ Immuno Standard Modules White and Black.

Features:

  • Available in 8-, 12- or 16-well strips
  • Framed or unframed for choice of optimal assay
  • Flat-bottom or C-shaped wells
  • Choice of three surface types for optimal binding
  • White format for highest possible signal reflection with low background fluorescence; recommended for epifluorescence
  • Black format to reduce backscatter light, which may be encountered in epifluorescence
  • Transparent or clear format for use in time-resolved fluorescence applications only

Choice of Surface:

  • Thermo Scientific™ PolySorp™ and Microlite™ 1+ are for adsorption of hydrophobic molecules.
  • Microlite 2+ is slightly hydrophilic and binds a diverse range of biomolecules.
  • Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assay.

WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Binding PropertyHydrophobic
Binding TypeAdsorption
BreakapartNo
CertifiedNo
Detection MethodFluorescence
For Use With (Application)Fluorescence
FormatStandard
No. per Case60
No. per Pack5
Packaging60 Cases
Plate BlockingNone
Product TypeMicroplate Strip
Quantity60/Cs.
Target MoleculeLipids
Volume (Metric) Well400 μL
Volume (Metric) Working350 μL
Array8 x 12
ColorBlack
MaterialPolystyrene
No. of Wells96
SterilityNon-sterile
Surface TreatmentPolySorp
Well ShapeRound, Flat Bottom (F-well)
Unit SizeCase of 60

Frequently asked questions (FAQs)

Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with all Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates?

Our Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules were designed to provide a positive seal for flat and round bottom wells of our Nunc MicroWell 96-Well Microplates and Nunc Immuno 96-Well Plates.
8-Well Strip Caps for Immuno Standard Modules are not compatible with C or V bottom wells of the Nunc Microwell 96-Well Microplates and Nunc Immuno 96-Well Plates.