CD3 Monoclonal Antibody (17A2), eFluor™ 450, eBioscience™ - FAQs
CD3 Monoclonal Antibody (17A2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (17A2), eFluor™ 450, eBioscience™ - FAQs (48-0032-80, 48-0032-82)