CD3 Monoclonal Antibody (OKT3), eFluor™ 450, eBioscience™ - FAQs
CD3 Monoclonal Antibody (OKT3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (OKT3), eFluor™ 450, eBioscience™ - FAQs (48-0037-41, 48-0037-42)