CD4 Monoclonal Antibody (SK3 (SK-3)), eFluor™ 450, eBioscience™ - FAQs
CD4 Monoclonal Antibody (SK3 (SK-3)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (SK3 (SK-3)), eFluor™ 450, eBioscience™ - FAQs (48-0047-41, 48-0047-42)