CD8a Monoclonal Antibody (OKT8 (OKT-8)), eFluor™ 450, eBioscience™ - FAQs
CD8a Monoclonal Antibody (OKT8 (OKT-8)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (OKT8 (OKT-8)), eFluor™ 450, eBioscience™ - FAQs (48-0086-41, 48-0086-42)