CD8a Monoclonal Antibody (SK1), eFluor™ 450, eBioscience™ - FAQs
CD8a Monoclonal Antibody (SK1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (SK1), eFluor™ 450, eBioscience™ - FAQs (48-0087-41)