CD14 Monoclonal Antibody (61D3), eFluor™ 450, eBioscience™ - FAQs
CD14 Monoclonal Antibody (61D3), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD14 Monoclonal Antibody (61D3), eFluor™ 450, eBioscience™ - FAQs (48-0149-41, 48-0149-42)