CD23 Monoclonal Antibody (B3B4), eFluor™ 450, eBioscience™ - FAQs
CD23 Monoclonal Antibody (B3B4), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD23 Monoclonal Antibody (B3B4), eFluor™ 450, eBioscience™ - FAQs (48-0232-80, 48-0232-82)