CD25 Monoclonal Antibody (PC61.5), eFluor™ 450, eBioscience™ - FAQs
CD25 Monoclonal Antibody (PC61.5), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (PC61.5), eFluor™ 450, eBioscience™ - FAQs (48-0251-82, 48-0251-80)