CD28 Monoclonal Antibody (CD28.2), eFluor™ 450, eBioscience™ - FAQs
CD28 Monoclonal Antibody (CD28.2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD28 Monoclonal Antibody (CD28.2), eFluor™ 450, eBioscience™ - FAQs (48-0289-42, 48-0289-41)