CD63 Monoclonal Antibody (H5C6), eFluor™ 450, eBioscience™ - FAQs
CD63 Monoclonal Antibody (H5C6), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD63 Monoclonal Antibody (H5C6), eFluor™ 450, eBioscience™ - FAQs (48-0639-41)