CD69 Monoclonal Antibody (FN50), eFluor™ 450, eBioscience™ - FAQs
CD69 Monoclonal Antibody (FN50), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD69 Monoclonal Antibody (FN50), eFluor™ 450, eBioscience™ - FAQs (48-0699-42, 48-0699-41)