CD73 Monoclonal Antibody (AD2), eFluor™ 450, eBioscience™ - FAQs
CD73 Monoclonal Antibody (AD2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD73 Monoclonal Antibody (AD2), eFluor™ 450, eBioscience™ - FAQs (48-0739-42, 48-0739-41)