Myeloperoxidase (MPO) Monoclonal Antibody (MPO455-8E6), eFluor™ 450, eBioscience™ - FAQs
Myeloperoxidase (MPO) Monoclonal Antibody (MPO455-8E6), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Myeloperoxidase (MPO) Monoclonal Antibody (MPO455-8E6), eFluor™ 450, eBioscience™ - FAQs (48-1299-42)