CD137 (4-1BB) Monoclonal Antibody (17B5), eFluor™ 450, eBioscience™ - FAQs
CD137 (4-1BB) Monoclonal Antibody (17B5), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD137 (4-1BB) Monoclonal Antibody (17B5), eFluor™ 450, eBioscience™ - FAQs (48-1371-82, 48-1371-80)