Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 450, eBioscience™ - FAQs
Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 450, eBioscience™ - FAQs (48-4031-80, 48-4031-82)