Ki-67 Monoclonal Antibody (SolA15), eFluor™ 450, eBioscience™ - FAQs
Ki-67 Monoclonal Antibody (SolA15), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Ki-67 Monoclonal Antibody (SolA15), eFluor™ 450, eBioscience™ - FAQs (48-5698-80, 48-5698-82)