FceR1 alpha Monoclonal Antibody (MAR-1), eFluor™ 450, eBioscience™ - FAQs
FceR1 alpha Monoclonal Antibody (MAR-1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for FceR1 alpha Monoclonal Antibody (MAR-1), eFluor™ 450, eBioscience™ - FAQs (48-5898-82, 48-5898-80)