Mouse IgD Monoclonal Antibody (11-26c (11-26)), eFluor™ 450, eBioscience™ - FAQs
Mouse IgD Monoclonal Antibody (11-26c (11-26)), eFluor™ 450, eBioscience™ - FAQs
View additional product information for Mouse IgD Monoclonal Antibody (11-26c (11-26)), eFluor™ 450, eBioscience™ - FAQs (48-5993-82, 48-5993-80)