IL-13 Monoclonal Antibody (eBio13A), eFluor™ 450, eBioscience™ - FAQs
IL-13 Monoclonal Antibody (eBio13A), eFluor™ 450, eBioscience™ - FAQs
View additional product information for IL-13 Monoclonal Antibody (eBio13A), eFluor™ 450, eBioscience™ - FAQs (48-7133-80, 48-7133-82)