CD45 Monoclonal Antibody (2D1), eFluor™ 450, eBioscience™ - FAQs
CD45 Monoclonal Antibody (2D1), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (2D1), eFluor™ 450, eBioscience™ - FAQs (48-9459-41, 48-9459-42)