HELIOS Monoclonal Antibody (22F6), eFluor™ 450, eBioscience™ - FAQs
HELIOS Monoclonal Antibody (22F6), eFluor™ 450, eBioscience™ - FAQs
View additional product information for HELIOS Monoclonal Antibody (22F6), eFluor™ 450, eBioscience™ - FAQs (48-9883-41, 48-9883-42)