TCR V beta 8 Monoclonal Antibody (JR-2), eFluor™ 450, eBioscience™ - FAQs
TCR V beta 8 Monoclonal Antibody (JR-2), eFluor™ 450, eBioscience™ - FAQs
View additional product information for TCR V beta 8 Monoclonal Antibody (JR-2), eFluor™ 450, eBioscience™ - FAQs (48-9890-41)