CD279 (PD-1) Monoclonal Antibody (J43), eFluor™ 450, eBioscience™ - FAQs
CD279 (PD-1) Monoclonal Antibody (J43), eFluor™ 450, eBioscience™ - FAQs
View additional product information for CD279 (PD-1) Monoclonal Antibody (J43), eFluor™ 450, eBioscience™ - FAQs (48-9985-80, 48-9985-82)