FDAA (Marfey's Reagent) (1-fluoro-2-4-dinitrophenyl-5-L-alanine amide)
Thermo Scientific™

FDAA (Marfey's Reagent) (1-fluoro-2-4-dinitrophenyl-5-L-alanine amide)

Thermo Scientific Pierce Marfey's Reagent (FDAA) reacts with primary amines to enable the quick and easy separation and quantitation ofRead more
Have Questions?
Catalog NumberQuantity
4889550 mg
Catalog number 48895
Price (USD)
136.00
Special offer
Online exclusive
Ends: 15-Jul-2026
160.00
Save 24.00 (15%)
Each
Quantity:
50 mg
Request bulk or custom format
Price (USD)
136.00
Special offer
Online exclusive
Ends: 15-Jul-2026
160.00
Save 24.00 (15%)
Each
Thermo Scientific Pierce Marfey's Reagent (FDAA) reacts with primary amines to enable the quick and easy separation and quantitation of optical isomers of amino acids by reverse-phase chromatography.

Features of Marfey's Reagent (FDAA):

• For precolumn derivatization of primary amines for detection by UV absorbance (λmax = 340nm)
• Provides nanomole sensitivity in typical HPLC applications with amino acids
• Reaction products enable chiral separation of D- and L-amino acids
• Following reverse-phase HPLC, derivatives have an extinction coefficient of 3 x 10^4M-1 cm-1

FDAA is 1-fluoro-2-4-dinitrophenyl-5-L-alanine amide, also called Marfey's reagent. The compound reacts with primary amines and is used as a derivatization reagent for UV detection in liquid chromatography methods. The reagent provides for detection at 340nm with nanomole sensitivity. FDAA derivatives of D-amino acids exhibit strong intramolecular bonding, which reduces their polarity relative to the corresponding L-amino acid derivatives. Consequently, the D-derivatives are selectively retained on reverse phase columns and elute much later than corresponding L-derivatives.

Reagent Properties:
• Alternative names: FDAA, Marfey's Reagent
• Chemical name: 1-fluoro-2-4-dinitrophenyl-5-L-alanine amide; N2-(5-Fluoro-2,4-dinitrophenyl)-L-alaninamide
• Reactive toward: Primary amines (—NH2)
• Chemical formula: C9H9FN4O5
• CAS number: 95713-52-3
• Molecular weight: 272.19
• Form: Bright yellow or orange crystals or powder, free of particulate matter

FDAA derivatives can be separated with simple linear gradient of triethylamine phosphate/acetonitrile on a Spheri-5 RP-18 HPLC cartridge column. FDAA reduces the need for a chiral column because the D-derivatives are retained on the column, eluting after the L-derivatives. The derivatization is easy to perform and completed in 90 minutes, resulting in derivatives that are stable for at least 48 hours.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Label or DyeOther Label or Dye
Quantity50 mg
FormSolid
Product TypeFDAA
Unit SizeEach
Contents & Storage
Upon receipt store product at room temperature.