MAGnify™ RARβ1 Primers

Catalog number: 492027

Applied Biosystems™  Related applications: Chromatin Biology | Real Time PCR (qPCR)

Error loading your content!

  Catalog number
Select a plan
Unit size
List price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Success in obtaining high-quality ChIP data is critically dependent on good primer design. The RARb1 primer set offer a sensitive and specific ChIP qPCR primer set compatible with SYBR® GreenER™ detection in a ready-to-use format. The R pre-made primer set has been functionally validated.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Species: Human
Gene Symbol: RARB
Target Gene: HAP, NR1B2, RRB2
Label or Dye: SYBR® GreenER™
Product Size: 100 reactions
Accession Number: U49855
Shipping Condition: Dry Ice

Contents & storage

Each Primer Set includes one vial of combined forward and reverse primers supplied in 100µl of DNase⁄RNAse free water at a 4µM concentration, sufficient for 100 20µl reactions. Store primers at -20°C in a non-frost-free freezer. Guaranteed stable for >6 months when properly stored.

Documents

Manuals & protocols