Premium Superfrost™ and Premium Plain Microscope Slides
 Related applications:
Anatomical Pathology
Achieve consistent results and facilitate interpretation with the Thermo Scientific™ Premium Superfrost™ and Premium Plain Slides, which are manufactured through a unique proprietary cleaning process. This process allows these slides to perform well in routine histology and microbiology applications, and specifically in hematology applications, by delivering consistently crisp, feathered-edge blood smears with good morphology.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
Premium Superfrost slides feature a smooth marking area for effortless writing, automated printing, or printed labels.
  • Manufactured from pure, water-white glass
  • Precleaned for immediate use
  • Superfrost slides feature a white ¾ in. tab
  • The premium Superfrost clipped-corner slide (Part No. 4951W-S-4-4370) is specifically designed for use with automated hematology instrumentation
Documents
Manuals & protocols