CaptureSelect™ Kappa XL Pre-packed Column

Catalog number: 494321005

 Related applications: Chromatography

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CaptureSelect™ KappaXL Affinity Matrix, available here in a pre-packed column format, has been specifically designed for the purification of recombinant human Ig’s (including IgA, IgM, IgD, and IgE) containing a Kappa light chain and their Fab fragments.

Features of this affinity matrix include:
• Mild elution for Fab fragments and antibodies
• Human specific (e.g., no binding to bovine antibodies)
• Excellent scalability
• Non-animal-derived

CaptureSelect™ KappaXL Affinity Matrix purifies Ig’s, Fab, and Fab2 fragments directly from complex source materials in a single step with high purity and yield.

Free of animal components
CaptureSelect™ products are affinity ligands created by a proprietary technology based on Camelid-derived single-domain antibody fragments. The ligand is a 13 kDa single-domain fragment comprising the 3 CDRs that form the antigen binding domain, efficiently produced by the yeast Saccharomyces cerevisiae in a production process free of animal components.

Characteristics:

Matrix: agarose-based, aldehyde-activated
Average particle size: 65 ± 10 µm
Ligand: CaptureSelect™ KappaXL affinity ligand
Coupling method: aldehyde coupling via NH2 residues of the ligand
Binding capacity: ~30 g IgG/L resin at 5-10 minutes contact time
Elution conditions: 20 mM citric acid, pH 3.5
Flow characteristics: 200–400 cm/h (up to 1 bar)
Formulation buffer: 20%(v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Affinity Column
Format: Chromatography Cartridge
Immobilized Phase: Anti-Kappa light chain
Product Line: CaptureSelect™
Product Size: 1 x 5 mL column
Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols