CD8a Monoclonal Antibody (AMC908), eFluor™ 660, eBioscience™ - FAQs
CD8a Monoclonal Antibody (AMC908), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (AMC908), eFluor™ 660, eBioscience™ - FAQs (50-0008-80, 50-0008-82)