CD25 Monoclonal Antibody (P4A10), eFluor™ 660, eBioscience™ - FAQs
CD25 Monoclonal Antibody (P4A10), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (P4A10), eFluor™ 660, eBioscience™ - FAQs (50-0250-42, 50-0250-41)