CD91 Monoclonal Antibody (A2MR-a2), eFluor™ 660, eBioscience™ - FAQs
View additional product information for CD91 Monoclonal Antibody (A2MR-a2), eFluor™ 660, eBioscience™ - FAQs (50-0919-42, 50-0919-41)