Mouse anti-Rat IgG2b Secondary Antibody, eFluor™ 660, eBioscience™ - FAQs
Mouse anti-Rat IgG2b Secondary Antibody, eFluor™ 660, eBioscience™ - FAQs
View additional product information for Mouse anti-Rat IgG2b Secondary Antibody, eFluor™ 660, eBioscience™ - FAQs (50-4815-82, 50-4815-80)