Osteopontin Monoclonal Antibody (2F10), eFluor™ 660, eBioscience™ - FAQs
Osteopontin Monoclonal Antibody (2F10), eFluor™ 660, eBioscience™ - FAQs
View additional product information for Osteopontin Monoclonal Antibody (2F10), eFluor™ 660, eBioscience™ - FAQs (50-9096-41, 50-9096-42)