EGFR Monoclonal Antibody (me1B3), eFluor™ 660, eBioscience™ - FAQs
EGFR Monoclonal Antibody (me1B3), eFluor™ 660, eBioscience™ - FAQs
View additional product information for EGFR Monoclonal Antibody (me1B3), eFluor™ 660, eBioscience™ - FAQs (50-9509-41, 50-9509-42)