IRF5 Monoclonal Antibody (ALYSCLN), eFluor™ 660, eBioscience™ - FAQs
View additional product information for IRF5 Monoclonal Antibody (ALYSCLN), eFluor™ 660, eBioscience™ - FAQs (50-9698-41, 50-9698-42)