Search
Search

| Número de catálogo | Capacidad (métrico) | Color del tapón | Modelo | Envase |
|---|---|---|---|---|
| 5000-0012GRN | 1.2 mL | Green | Cryobox 9 x 9 | 500 Tubes |
| 5000-1012RED | 1 mL | Red | Cryobox 9 x 9 | 500 Tubes |
| 5000-1012ORG | 1 mL | Orange | Cryobox 9 x 9 | 500 Tubes |
| 5000-1012YEL | 1 mL | Yellow | Cryobox 9 x 9 | 500 Tubes |
| 5000-1012GRN | 1 mL | Green | Cryobox 9 x 9 | 500 Tubes |
| 5000-1012BLU | 1 mL | Blue | Cryobox 9 x 9 | 500 Tubes |
| 5000-1012PUR | 1 mL | Purple | Cryobox 9 x 9 | 500 Tubes |
| 5000-1020RED | 1.5 mL | Red | 10 x10 System 100 | 500 Tubes |
| 5000-1020ORG | 1.5 mL | Orange | 10 x10 System 100 | 500 Tubes |
| 5000-1020YEL | 1.5 mL | Yellow | 10 x10 System 100 | 500 Tubes |
| 5000-1020GRN | 1.5 mL | Green | 10 x10 System 100 | 500 Tubes |
| 5000-1020BLU | 1.5 mL | Blue | 10 x10 System 100 | 500 Tubes |
| 5000-1020PUR | 1.5 mL | Purple | 10 x10 System 100 | 500 Tubes |
| 5000-0012RED | 1.2 mL | Red | Cryobox 9 x 9 | 500 Tubes |
| 5000-0012ORG | 1.2 mL | Orange | Cryobox 9 x 9 | 500 Tubes |
| 5000-0012YEL | 1.2 mL | Yellow | Cryobox 9 x 9 | 500 Tubes |
| 5000-0012BLU | 1.2 mL | Blue | Cryobox 9 x 9 | 500 Tubes |
| 5000-0012PUR | 1.2 mL | Purple | Cryobox 9 x 9 | 500 Tubes |
| 5012-0012RED | 1.2 mL | Red | Cryobox 9 x 9 | 1000 Tubes |
| 5012-0012ORG | 1.2 mL | Orange | Cryobox 9 x 9 | 1000 Tubes |
| 5012-0012YEL | 1.2 mL | Yellow | Cryobox 9 x 9 | 1000 Tubes |
| 5012-0012GRN | 1.2 mL | Green | Cryobox 9 x 9 | 1000 Tubes |
| 5012-0012BLU | 1.2 mL | Blue | Cryobox 9 x 9 | 1000 Tubes |
| 5012-0012PUR | 1.2 mL | Purple | Cryobox 9 x 9 | 1000 Tubes |
| 5010-0020RED | 2 mL | Red | Cryobox 9 x 9 | 500 Tubes |
| 5010-0020ORG | 2 mL | Orange | Cryobox 9 x 9 | 500 Tubes |
| 5010-0020YEL | 2 mL | Yellow | Cryobox 9 x 9 | 500 Tubes |
| 5010-0020GRN | 2 mL | Green | Cryobox 9 x 9 | 500 Tubes |
| 5010-0020BLU | 2 mL | Blue | Cryobox 9 x 9 | 500 Tubes |
| 5010-0020PUR | 2 mL | Purple | Cryobox 9 x 9 | 500 Tubes |
| 5012-0020RED | 2 mL | Red | Cryobox 9 x 9 | 1000 Tubes |
| 5012-0020ORG | 2 mL | Orange | Cryobox 9 x 9 | 1000 Tubes |
| 5012-0020YEL | 2 mL | Yellow | Cryobox 9 x 9 | 1000 Tubes |
| 5012-0020GRN | 2 mL | Green | Cryobox 9 x 9 | 1000 Tubes |
| 5012-0020BLU | 2 mL | Blue | Cryobox 9 x 9 | 1000 Tubes |
| 5012-0020PUR | 2 mL | Purple | Cryobox 9 x 9 | 1000 Tubes |
| 5010-0050RED | 5 mL | Red | Cryobox 9 x 9 | 250 Tubes |
| 5010-0050ORG | 5 mL | Orange | Cryobox 9 x 9 | 250 Tubes |
| 5010-0050YEL | 5 mL | Yellow | Cryobox 9 x 9 | 250 Tubes |
| 5010-0050GRN | 5 mL | Green | Cryobox 9 x 9 | 250 Tubes |
| 5010-0050BLU | 5 mL | Blue | Cryobox 9 x 9 | 250 Tubes |
| 5010-0050PUR | 5 mL | Purple | Cryobox 9 x 9 | 250 Tubes |
Features: