This product will no longer be available on thermofisher.com after Dec. 28, 2020. Learn more »
Thermo Scientific™
Richard-Allan Scientific™ Mark-It™ Tissue Marking Dyes
Related applications:
Anatomical Pathology
For a variety of tissue identification purposes use Thermo Scientific™ Richard-Allan Scientific™ Mark-It™ Tissue Marking Dyes.
Back to top
Features:
  • Permanent dyes that maintain their original brilliance during routine tissue processing cycles
  • Can be used on fresh or formalin-fixed tissues
Uses include:
  • Marking surgical margins
  • Identifying serial segments of large tissues to ensure proper orientation and embedding
  • Maintaining proper tissue orientation for Mohs surgery specimens
  • Identification of multiple pieces of tissues processed in the same cassette
Ordering Information:
  • Available in black, blue, green, orange, red and yellow in 2 oz. bottles